Dropper® Spinal Fluid Control, Quantimetrix
Supplier: Quantimetrix
Certificates
About this item
Dropper Spinal Fluid Control monitors the performance of spinal fluid chemistry assay methods. Drop the control directly into the instrument sample cup with an easy-to-use, dropper bottle for simple dispensing and maximum use. Dropper Spinal Fluid Control is designed for use with most major chemistry analyzers and nephelometers including Beckman Coulter Synchron®, Siemens Dimension®, Beckman Coulter AU® series, Ortho Vitros®, Roche Cobas®, Abbott Arcitect and Pyrogallol Red microprotein methods.
- Easy-to-use dropper bottles
- Liquid, human protein matrix
- 2-year refrigerated shelf life from date of manufacture
- 90-day open vial stability when stored at 2 to 8°C
Ordering information: Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.
Frequently Bought Together