Bilirubin Control® Pediatric, Quantimetrix
Supplier: Quantimetrix
Certificates
About this item
Use Bilirubin Control – Pediatric to monitor the performance of neonatal bilirubin assay methods including Reichert Unistat® and Advanced® Instruments BR2 Bilirubin Stat-Analyzer™ Model bilirubinometers and all automated bilirubin methods for popular instruments including Abbott Architect, Beckman Coulter Synchron®, Beckman Coulter AU Series, Ortho Vitros®, Roche Modular®, Roche Cobas®, Siemens Advia® and Siemens Dimension®.
- Neonatal bilirubin levels up to 20 mg/dL
- Liquid, human protein matrix
- 2-year refrigerated shelf life from date of manufacture
- 5-day open vial stability when stored at 2 to 8°C (35 to 46°F)
Ordering information: Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.
Frequently Bought Together