Order Entry
United States
ContactUsLinkComponent
Paper Labels for 3" Core Printers
Paper Labels for 3" Core Printers
Catalog # 77532-232
Supplier:  Brady Worldwide
Paper Labels for 3" Core Printers
Catalog # 77532-232
Supplier:  Brady Worldwide
Supplier Number:  30240

Specifications

  • For Use With:
    BBP81, i5100, i7100, IP Series, PR Plus, THT 3" core printers
  • Color:
    White
  • Description:
    Paper labels for direct thermal printers, rectangular
  • Dimensions:
    0.5H×2"W
  • Material:
    Paper
  • Thickness:
    3.5 mil.
  • Cat. No.:
    77532-232
  • Packaging:
    3000 labels/roll
  • Core size:
    3"

Specifications

About this item

General purpose paper with matte finish and smooth top coat finish for thermal transfer printing.

  • White matte paper label stock
  • Permanent rubber adhesive
  • Low cost is ideal for all general purpose labeling projects

Ideal for use in general labeling and barcoding applications where the label needs to be removed or repositionable. A ribbon is required to print this material.