Order Entry
United States
ContactUsLinkComponent
DHEAS ELISA
DHEAS ELISA
Catalog # 101720-136
Supplier:  Alpco Diagnostics
CAS Number:  
DHEAS ELISA
Catalog # 101720-136
Supplier:  Alpco Diagnostics
Supplier Number:  11-DHEHU-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    DHEA-S
  • Description:
    DHEAS ELISA
  • Size:
    1×96 tests
  • Sample Type:
    Serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h
  • Detection Range:
    0.005 - 10 µg/ml
  • Sensitivity:
    5 ng/ml
  • Regulatory Status:
    FDA registered. For in vitro diagnostic use.
  • Cat. No.:
    101720-136
  • Supplier no.:
    11-DHEHU-E01

Specifications

About this item

The Dehydroepiandrosterone Sulfate ELISA is intended for the direct quantitative determination of DHEA-S by enzyme immunoassay in human serum.

Dehydroepiandrosterone sulfate (DHEAS) is produced by the adrenals and gonads. As a result, the determination of the level of DHEAS in serum is important in the evaluation of the functional state of these glands. DHEAS is a precursor of testosterone and estrone. Besides the adrenals in females, the ovaries have been shown to be an important source of DHEAS. It has been reported that there is a fluctuation day by day of DHEAS in women during the ovulatory cycle. The principle production of testosterone in females is from conversion of other related androgens, especially DHEAS. An abnormal testosterone level in women should be accompanied by the estimation of serum DHEAS. The use of serum testosterone determination in conjunction with the ELISA of DHEAS can be used to determine if the source of excess androgen production is ovarian or adrenal.

Certifications: FDA Registered.
Caution: For In Vitro diagnostic use.