Order Entry
United States
ContactUsLinkComponent
Progesterone ELISA
Progesterone ELISA
Catalog # 101720-142
Supplier:  Alpco Diagnostics
CAS Number:  
Progesterone ELISA
Catalog # 101720-142
Supplier:  Alpco Diagnostics
Supplier Number:  11-PROHU-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Progesterone
  • Description:
    Progesterone ELISA
  • Size:
    1×96 tests
  • Sample Type:
    serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h, 10 min
  • Detection Range:
    0.3 - 60 ng/ml
  • Sensitivity:
    0.1 ng/ml
  • Regulatory Status:
    FDA registered. For in vitro diagnostic use.
  • Cat. No.:
    101720-142
  • Supplier no.:
    11-PROHU-E01

Specifications

About this item

The Progesterone ELISA is for the direct quantitative determination of Progesterone by enzyme immunoassay in human Serum.

  • FDA registered; for in vitro diagnostic use

Progesterone is a C-21 female sex steroid hormone with a variety of physiological effects. In the follicular phase of the menstrual cycle, progesterone is produced in low levels. It increases to the LH peak and then sharply rises 3 to 4 days later to higher levels, remaining elevated through the 10th to 12th days after the LH peak. Then there is a sharp decline to the low levels of the follicular phase. It is responsible for the induction of the cyclic changes in the endometrium of the uterus allowing implantation and successful growth of the fertilized ovum and maintenance of pregnancy.

Progesterone measurements are used in documenting ovulation and in the management of difficulties during the first trimester of pregnancy. Levels of progesterone may be useful in the evaluation of sterility due to luteal phase defects, prediction of impending abortion, and the diagnosis of ectopic pregnancy. Normal values of progesterone may be affected by drugs such as, oral contraceptives, super ovulatory drugs, estrogen replacement therapy medication, and GnRH analogues. The removal of ovarian function following surgical oophorectomy or chemotherapy may influence serum progesterone values.

Certifications: FDA registered. For in vitro diagnostic use in the United States.