BBL™ Trypticase™ Soy Agar (IC-XT) Plates
Catalog # 76452-424
Supplier: BD
CAS Number:
Specifications
- Sterility:Sterile
- Cat. No.:76452-424
- Plate diameter:100 mm
Specifications
About this item
BD Isolator Cleanroom eXtended Temperature (IC-XT) plated media offer flexibility and sample security to meet today's environmental monitoring needs.
- Sterility assurance
- VHP resistance
- Sample integrity and product traceability
- Storage flexibility and resource efficiency
- Multiple applications
- Air sampler compatibility
With extended shelf-life and 2 to 25 °C storage, BD BBL™ IC-XT prepared plated media provide dependable and reliable products for testing in critical environments.