ContactUsLinkComponent
Acrodisc™ Syringe Filters with PTFE Membrane, Cytiva (Formerly Pall Lab)
Acrodisc™ Syringe Filters with PTFE Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Certificates
undefined
Acrodisc™ Syringe Filters with PTFE Membrane, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
New Product
Certificates
Specifications
  • Catalog No:
  • 77553-284
  • 77553-288
  • 77553-286
  • 77553-314
  • 77553-296
  • 77553-312
  • Type:
  • 77553-284
  • 77553-288
  • 77553-286
  • 77553-314
  • 77553-296
  • 77553-312
  • Hold-up Volume:
  • <125 µl
  • <125 µl
  • <125 µl
  • <125 µl
  • <14 µl
  • <125 µl
  • Max. Operating Pressure:
  • 30.4 psi
  • 30.4 psi
  • 59.4 psi
  • 30.4 psi
  • 100 psi
  • 30.4 psi
  • Max. Temperature:
  • 100 °C
  • 100 °C
  • 55 °C
  • 100 °C
  • 100 °C
  • 100 °C
  • Water Flow Rate:
  • 395 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 510 ml/min at 2.1 bar (210 kPa, 30 psi)
  • 215 ml/min at 2.1 bar (210 kPa, 30 psi)
  • 510 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 510 ml/min at 1.0 bar (100 kPa, 15 psi)
  • 245 ml/min at 1.0 bar (100 kPa, 15 psi)