Order Entry
United States
ContactUsLinkComponent
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanamine ≥98%
Corrosive
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanamine ≥98%
Catalog # 77884-514
Supplier:  Ambeed
New Product
CAS Number:  
Corrosive
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanamine ≥98%
Catalog # 77884-514
Supplier:  Ambeed
Supplier Number:  A649987-100MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

About this item

 :161.2 g/mol
UN:3259
ADR:8,Ⅲ
 :GHS05
CAS Number:4649-12-1
 :
Formula:C₉H₁₁N₃
MDL Number:MFCD10697517