Sodium taurocholate hydrate ≥97%
New Product
Supplier: Ambeed
Jump to:
About this item
Caution: for Research Use Only
Synonyms: Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate
- CAS Number:345909-26-4
- Formula:C₂₆H₄₆NNaO₈S
- MDL Number:MFCD00150819
- Molecular Weight:555.70 g/mol
4 Options Available Below
Product Details & Documents
Caution: for Research Use Only
Synonyms: Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate
- CAS Number:345909-26-4
- Formula:C₂₆H₄₆NNaO₈S
- MDL Number:MFCD00150819
- Molecular Weight:555.70 g/mol
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A555499-25GSpecifications:New ProductNew Product
- Supplier Number: A555499-50MG
- Supplier Number: A555499-250MGSpecifications:New ProductNew Product
- Glass bottle1 gDiscontinued-Supplier Number: A555499-1GSpecifications:New ProductNew Product
- Supplier Number: A555499-5GSpecifications:New ProductNew Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



