Order Entry
United States
ContactUsLinkComponent
Ethyl 3-hydrazinylbenzoate hydrochloride ≥95%, Ambeed.Inc
Irritant
Ethyl 3-hydrazinylbenzoate hydrochloride ≥95%, Ambeed.Inc
Catalog # 77683-711
Supplier:  Ambeed
New Product
CAS Number:  
Irritant
Ethyl 3-hydrazinylbenzoate hydrochloride ≥95%, Ambeed.Inc
Catalog # 77683-711
Supplier:  Ambeed
Supplier Number:  A238070-5G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g
  • Cat. No.:
    77683-711

Specifications

About this item

CAS Number:940875-99-0
 :
Formula:C₉H₁₃ClN₂O₂
MDL Number:MFCD06796173
 :216.66 g/mol
 :GHS07