Calibration weights, OIML M1
Supplier: Mettler Toledo
Certificates
About this item
Set M1 OIML class weight in a plastic box including accessories for handling and cleaning and a calibration certificate.
- Stainless steel weights
Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.
Frequently Bought Together