Membrane Disc Filters, GLA-5000, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
Ideal for sampling airborne metals, silica, and dust.
- Polyvinyl chloride
- Resistant to acids and alkalis
- For gravimetric analysis
Polyvinyl chloride (PVC) membranes resist acids and alkalis. Low ash content, moisture pick-up, and tare weight make membranes well suited for gravimetric analysis of airborne particulates. Pore size: 5 µm. Filter 28139-743 includes support pads.
Certifications: Meets NIOSH and OSHA requirements.