Order Entry
United States
ContactUsLinkComponent
Steel Closed Head Container, UN Marked, Qorpak®
Steel Closed Head Container, UN Marked, Qorpak®
Catalog # 16465-019
Supplier:  Berlin Packaging
Small Business Enterprise
undefined
Steel Closed Head Container, UN Marked, Qorpak®
Catalog # 16465-019
Supplier:  Berlin Packaging
Supplier Number:  HMS-6741M
Small Business Enterprise

Specifications

  • Description:
    Steel closed head container
  • Cat. No.:
    16465-019
  • Supplier no.:
    HMS-6741M

Specifications

About this item

One-gallon steel closed head drum is suspended in a die-cut carrier.

  • Unlined interior
  • Complete with plastic bag, tape and instructions
  • Pressure capability of 250 kPa

Acceptable for Great Danger Hazardous Materials (gross mass not exceeding 9.5 kg) Packing Group I.