ContactUsLinkComponent
MOPS Minimal Medium Kit, Teknova
MOPS Minimal Medium Kit, Teknova
Catalog # 100218-936
Supplier:  Teknova
CAS Number:  
MOPS Minimal Medium Kit, Teknova
Catalog # 100218-936
Supplier:  Teknova
Supplier Number:  M2106
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    MOPS Minimal Medium Kit
  • Cat. No.:
    100218-936
  • Supplier no.:
    M2106
Specifications
About this item
Designed for the growth of E
MOPS mixture should be stored frozen. The potassium phosphate dibasic solution and 20% glucose solution should be stored at room temperature.
coli and other enterobacteria. Kit consists of 100mL 10X MOPS mixture (200059-678), 10mL 0.132M potassium phosphate diabase solution (200059-652), and 100mL 20% glucose solution (100216-728), and makes 5 liters.