Order Entry
United States
ContactUsLinkComponent
Kovac's indole reagent for microbiology ph Value: <1 (H₂O, 20 °C), Millipore®
Kovac's indole reagent for microbiology ph Value: <1 (H₂O, 20 °C), Millipore®
Catalog # EM1.09293.0100
Supplier:  MilliporeSigma
CAS Number:  
Kovac's indole reagent for microbiology ph Value: <1 (H₂O, 20 °C), Millipore®
Catalog # EM1.09293.0100
Supplier:  MilliporeSigma
Supplier Number:  1.09293.0100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    EM1.09293.0100
  • Supplier no.:
    1.09293.0100

Specifications

Specification Test Results

  • Indole formation (Escherichia coli ATCC 25922 (WDCM 00013)):
    +
  • Indole formation (Proteus vulgaris ATCC 8427):
    +
  • Indole formation (Enterobacter aerogenes ATCC 13048):
    -

Specification Test Results

About this item

Synonyms: 4-(Dimethylamino)benzaldehyde solution
 :
 :149.19 g/mol
Storage Temperature:Refrigerator
UN:2920
ADR:8 (3),II
 :GHS02
 :GHS05
 :GHS07
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381