Order Entry
United States
ContactUsLinkComponent
 
Lab Pack, UN and DOT, Eagle Manufacturing
  56617-331
undefined
Lab Pack, UN and DOT, Eagle Manufacturing
  56617-331
 :  1652

 

  • Dimensions:
    45.7O.D.×56.5H cm (18×221/4")
  • Cat. No.:
    56617-331
  • Weight:
    4 kg (9 lbs.)
  • Supplier no.:
    1652

 

 

Suitable for use with hazardous materials, Packing Groups I, II, and III.

Constructed of blow-molded high-density polyethylene with UV inhibitors. Chemical-resistant to most materials. Incinerates completely. Nestable for easy storage. Handles are suitable for forklifting.

UN certified to HM-181. Meets DOT 49 CFR 173.12(b). UN Marking: 1H2/x40/S.

Holds 18.9L (5gal.) pails or smaller.