Mouse Recombinant TNFRSF18 (ECD, His Tag) (from HEK293 cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse TNFRSF18 (NP_033426.1) (Ser22-Gln150) was expressed with a polyhistidine tag at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:ECD, His Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 °C. Store it under sterile conditions at −20 to −80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Y
- Protein Synonyms:Gitr|AITR
- Protein/Peptide Name:TNFRSF18
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse TNFRSF18 consists of 140 amino acids and predicts a molecular mass of 15.5 kDa.
- Sequence:Ser22-Gln150
- Formulation:Lyophilized from sterile PBS, pH 7.4.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together