Streptococcus pyogenes Recombinant SD90_RS03345 (His and GST Tag) (from E. coli)
77444-516
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Streptococcus pyogenes
- Size:20 kU
- Tag sequence:His and GST Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 °C. Store it under sterile conditions at −20 to −80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Biological Activity:One unit digests ≥95% of 1 µg human IgG when incubated in PBS buffer, pH 7.4 at 37°C for 30min. The specific activity is 20,000 unit/mg.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Y
- Protein/Peptide Name:SD90_RS03345
- Purity:>85% as determined by SDS-PAGE.
- Molecular Weight:The recombinant streptococcus pyogenes IdeS consists of 549 amino acids and predicts a molecular mass of 63.1 kDa.
- Sequence:Asp30-Asn341
- Formulation:Supplied as sterile PBS, 50% glycerol, pH 7.4.
- Cat. No.:77444-516
A DNA sequence encoding the streptococcus pyogenes IdeS (F8V4V0-1) (Asp30-Asn341) was expressed with the N-terminal polyhistidine-tagged a GST tag at the N-terminus (his-GST)
- Shipping temperature: Dry ice