Edge–Rite™ Disposable Microtome Blades, Epredia™, Richard-Allan Scientific
Supplier: Avantor
Certificates
About this item
Experience consistent quality from blade to blade and sample to sample with Epredia™ Edge–Rite™ Disposable Microtome Blades.
- Surgical–grade steel
- High– and low–profile blades are specially coated
- Increase longevity and optimize section quality
- Low profile blades are 20% thicker than traditional blades
- Increase knife edge stability
- Allows for excellent ribboning and facilitates usage in cryostats
- Blades are 5% longer
- Allowing for more sections per blade
- Blades fit any low–profile blade holder
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.