ContactUsLinkComponent
InLab® Redox Pro Combination Electrode, Sleeve Junction, METTLER TOLEDO®
InLab® Redox Pro Combination Electrode, Sleeve Junction, METTLER TOLEDO®
Catalog # 97001-692
Supplier:  Mettler Toledo
Product available on GSA Advantage®
undefined
InLab® Redox Pro Combination Electrode, Sleeve Junction, METTLER TOLEDO®
Catalog # 97001-692
Supplier:  Mettler Toledo
Supplier Number:  51343201
Product available on GSA Advantage®
Specifications
  • Temperature range:
    0...100 °C
  • Length:
    120 mm
  • Material:
    Glass
  • Cat. No.:
    97001-692
  • Internal reference:
    ARGENTHAL™
  • Junction:
    Sleeve
  • Supplier no.:
    51343201
  • Electrolyte:
    3M KCl
Specifications
About this item
Electrodes measure oxidation/reduction power of a solution
  • Platinum ring indicator
  • Moveable glass sleeve
  • Safelock™ closing ring
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head connector
They feature a platinum ring indicator, a moveable glass sleeve, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.