Order Entry
United States
ContactUsLinkComponent
 
InLab® Mono Pro pH Half-Cell Electrode, METTLER TOLEDO®
  97001-672
Product available on GSA Advantage®
undefined
InLab® Mono Pro pH Half-Cell Electrode, METTLER TOLEDO®
  97001-672
 :  51343196
Product available on GSA Advantage®

 

  • Temperature range:
    0...130 °C
  • Length:
    120 mm
  • Ø:
    12 mm
  • Cat. No.:
    97001-672
  • pH range:
    0 - 12
  • Body material:
    Glass
  • Junction:
    Ceramic
  • Supplier no.:
    51343196

 

 

Electrode is recommended for applications in which the lifetime of the electrode is significantly shorter than that of a standard reference electrode

  • Thick-walled A41 glass body
  • S7 screw head connector

Thick-walled A41 glass body provides extra protection against corrosive and organic samples and is ideal for high temperatures and biological media. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.