RSV Recombinant RSV-F (from CHO Cells), Biotinylated
Catalog # 77443-980
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:RSV
- Size:20 µg
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at –20 ℃...–80 ℃. Store it under sterile conditions at –20 ℃...–80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Immobilized anti-RSV prefuison F protein antibody which identifies Site Ø epitope at 2 μg/ml (100 μl/well) can bind Recombinant RSV(A2)Prefusion glycoprotein F/ RSV-preF Protein,Biotinylated (Cat: 11049-VNAS-B), the EC50 is 1.8 - 5.4 ng/ml.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Y
- Protein Synonyms:HRSVgp08|F
- Protein/Peptide Name:RSV-F
- Purity:≥90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant RSV (A2)Prefusion glycoprotein F has a predicted molecular mass of 54.5 kDa.It migrates as an approximately 59.9 kDa band in SDS-PAGE under reducing conditions.
- Formulation:Lyophilized from sterile PBS, pH 7.4.
- Shipping Temperature:Ambient
- Cat. No.:77443-980