ContactUsLinkComponent
STEMdiff™ Sensory Neuron Differentiation Kit
STEMdiff™ Sensory Neuron Differentiation Kit
Catalog # MSPP-100-0341
CAS Number:  
undefined
STEMdiff™ Sensory Neuron Differentiation Kit
Catalog # MSPP-100-0341
Supplier Number:  100-0341
CAS Number:  
Specifications
  • Description:
    STEMdiff™ Sensory neuron differentiation kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-100-0341
Specifications
About this item
Differentiation kit for the generation of sensory neuronal precursors from human iPS cell-derived neural crest cells.
Generate functional, human-specific sensory neurons in a dish with this easy-to-use culture system. After obtaining pluripotent stem cell (PSC)-derived neural crest cells (NCCs) with STEMdiff™ Neural Crest Differentiation Kit, use the serum-free STEMdiff™ Sensory Neuron Differentiation Kit to generate sensory neuron precursors. STEMdiff™ Sensory Neuron Maturation Kit can then be used to maintain and mature the sensory neuron precursors.
The resulting cells are BRN3A-positive and more than 70% TUJ1-positive. With the physiological glucose and osmolarity conditions provided by BrainPhys™ as the basal medium, the neurons exhibit activity in response to sensory ligands and changes in temperature. The generated population of functional human sensory neurons can be used for drug discovery and pain research applications.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').