StemSpan™ ACF Erythroid Expansion Medium
Catalog # MSPP-09860
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Description:StemSpan™-ACF erythroid expansion medium
- Size:500 ml
- Cat. No.:MSPP-09860
Specifications
About this item
Animal component-free medium for culture and expansion of human erythroid cells.
StemSpan™-ACF Erythroid Expansion Medium (ACF-E) is an animal component-free (ACF) medium for the in vitro culture and expansion of human hematopoietic cells. When supplemented with StemSpan™ Erythroid Expansion Supplement 100X, this medium supports the expansion and erythroid differentiation of human hematopoietic progenitor cells into CD71+ and Glycophorin A+ erythroblasts in ACF culture conditions. This medium only contains recombinant proteins and synthetic components; it does not contain serum or other human- or animal-derived components.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').