ContactUsLinkComponent
MEM Non-Essential Amino Acid Solution (100X)
MEM Non-Essential Amino Acid Solution (100X)
Catalog # MSPP-07600
CAS Number:  
undefined
MEM Non-Essential Amino Acid Solution (100X)
Catalog # MSPP-07600
Supplier Number:  07600
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    MEM Non-Essential amino acid solution (100X)
  • Size:
    100 ml
  • Cat. No.:
    MSPP-07600
Specifications
About this item
MEM non-essential amino acid solution (100X).
MEM Non-Essential Amino Acid Solution (100X) has been pre-screened for use with other reagents of the ES-Cult™ product line for optimal performance for the in vitro hematopoietic differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').