N2 Supplement-A
Catalog # MSPP-07152
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Description:N2 Supplement-A
- Size:5 ml
- Cat. No.:MSPP-07152
Specifications
About this item
For neural and pancreatic differentiation of mouse and human ES and iPS cells.
N2 Supplement-A, containing iron-rich human transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to neural and pancreatic-like cell types. Different neuronal subtypes can be generated when human ES/iPS cell-derived neural progenitor cells are cultured in BrainPhys™ Neuronal Medium supplemented with N2 Supplement-A, NeuroCult™ SM1 Neuronal Supplement, and other factors. N2 Supplement-A is provided as a 100X stock solution. N2 Supplement-A is available for individual sale or as a component of the BrainPhys™ Neuronal Medium N2-A & SM1 Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').