- Description:NeuroCult™ Proliferation kit (Mouse & Rat)
- Size:500 ml
- Cat. No.:MSPP-05702
Medium kit for expansion of mouse and rat neural stem and progenitor cells.
NeuroCult™ Proliferation Kit (Mouse and Rat) is a standardized, serum-free medium and supplement kit for the culture of mouse and rat neural stem and progenitor cells from normal tissues or tumor samples. When supplemented with appropriate cytokines, NeuroCult™ Proliferation Kit (Mouse and Rat) is optimized to maintain mouse and rat neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential.
NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF is required. When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF and Heparin Solution are also required.