ContactUsLinkComponent
MesenCult™ Expansion Kit (Mouse)
MesenCult™ Expansion Kit (Mouse)
Catalog # MSPP-05513
CAS Number:  
undefined
MesenCult™ Expansion Kit (Mouse)
Catalog # MSPP-05513
Supplier Number:  05513
CAS Number:  
Specifications
  • Description:
    MesenCult™ Expansion kit (Mouse)
  • Size:
    1 kit
  • Cat. No.:
    MSPP-05513
Specifications
About this item
For the culture of mouse MSCs and MEFs.
MesenCult™ Expansion Kit (Mouse) is standardized for the culture of mouse mesenchymal stromal cells (MSCs; also known as mesenchymal stem cells) and mouse embryonic fibroblasts (MEFs). The kit includes MesenCult™ Basal Medium (Mouse), MesenCult™ 10X Supplement (Mouse), and MesenPure™. MesenCult™ Expansion Medium has been optimized for the derivation and expansion of mouse MSCs and MEFs in vitro as well as for the detection of colony-forming unit–fibroblasts (CFU­F). This kit was optimized using cells from the mouse strain C57BL/6. To facilitate the enrichment of MSCs and MEFs during cell culture without serial passaging and frequent medium changes, simply add MesenPure™ to complete MesenCult™ Expansion Medium just prior to use. Although not required, the addition of MesenPure™ is strongly recommended, as the resulting MSC and MEF cultures are more homogeneous and exhibit more robust proliferation, differentiation, and colony formation when compared to complete MesenCult™ Expansion Medium alone.
NOTE: MesenCult™ Expansion Medium must be supplemented with L-Glutamine.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').