ContactUsLinkComponent
MegaCult™ C Collagen and Medium without Cytokines
MegaCult™ C Collagen and Medium without Cytokines
Catalog # MSPP-04960
CAS Number:  
undefined
MegaCult™ C Collagen and Medium without Cytokines
Catalog # MSPP-04960
Supplier Number:  04960
CAS Number:  
Specifications
  • Description:
    MegaCult™-C Collagen and medium without cytokines
  • Size:
    1 kit
  • Cat. No.:
    MSPP-04960
Specifications
About this item
Kit containing collagen and medium without cytokines for human and mouse CFU-Mk assays.
MegaCult™-C Collagen and Medium without Cytokines kit includes medium and collagen solution necessary for performing colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk). MegaCult™-C Medium Without Cytokines is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples, after addition of appropriate cytokines.
It is suitable for use with CD34 + enriched cells, mononuclear cells, and cells isolated by other purification methods. It is also intended for assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow, after addition of appropriate cytokines. Collagen Solution for preparation of collagen gels and for coating cell culture surfaces. Individual kit components may also be purchased separately, for your convenience. For more information on protocols for human CFU-Mk assays using MegaCult™-C, please explore the technical manual.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').