MegaCult™ C Medium with Lipids
Catalog # MSPP-04850
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Description:MegaCult™-C medium with lipids
- Size:50 ml
- Cat. No.:MSPP-04850
Specifications
About this item
Medium with lipids, without cytokines.
MegaCult™-C Medium with Lipids is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples after addition of appropriate cytokines. It is recommended for use with CD34+-enriched cells, mononuclear cells, and cells isolated by other purification methods. It can also be used for CFU-MK assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow after addition of appropriate cytokines.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').