DMEM/F-12 with 15 mm HEPES
Catalog # MSPP-36254
Supplier: Stemcell Technologies
CAS Number:
Specifications
- Description:DMEM/F-12 with 15 mm HEPES
- Size:500 ml
- Cat. No.:MSPP-36254
Specifications
About this item
Dulbecco's Modified Eagle's Medium/Nutrient Ham's Mixture F-12 (DMEM/F-12) with 15 mm HEPES buffer.
Dulbecco's Modified Eagle's Medium/Nutrient Ham's Mixture F-12 (DMEM/F-12) with 15 mm HEPES is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on cell type, culture conditions, and degree of chemical definition required for the cell culture application. This product has been pre-screened for use with other reagents of the ES-Cult™ product line for maintenance culture of embryonic stem (ES) cells in the undifferentiated state.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').