Vesta-Bac HB Ready-to-Use (RTU) One-Step Disinfectant
76531-304
: Steris
- Size:22 oz.
- Cat. No.:76531-304
- Packaging:Plastic bottle (12/case)
Vesta-Bac HB Disinfectant is a ready-to-use liquid specifically formulated for the disinfection of hard surfaces. The product can be sprayed directly on surfaces.
- Efficacy - Broad spectrum activity including virucidal activity against HBV, HCV, and HIV-1 (associated with AIDS)
- Productivity - Ready-to-use formulation saves time and labor
- Flexibility - Cleans, deodorizes, disinfects and can also be used to control mold and mildew
- Reliability - Bactericidal, fungicidal and virucidal in the presence of organic soil (5 percent bovine serum)
Vesta-Bac HB Disinfectant is a ready-to-use one-step disinfectant that has a broad range of activity against gram-negative and gram-positive bacteria, viruses including Hepatitis B (HBV), Hepatitis C (HCV), HIV-1, and fungi.