ContactUsLinkComponent
STEMdiff™ Endothelial Expansion Medium Kit
STEMdiff™ Endothelial Expansion Medium Kit
Catalog # MSPP-08007
CAS Number:  
undefined
STEMdiff™ Endothelial Expansion Medium Kit
Catalog # MSPP-08007
Supplier Number:  08007
CAS Number:  
Specifications
  • Description:
    STEMdiff™ Endothelial expansion medium kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-08007
Specifications
About this item
Serum- and xeno-free medium for expansion of human pluripotent stem cell-derived endothelial cells.
STEMdiff™ Endothelial Expansion Medium Kit includes the xeno-free endothelial expansion basal medium and supplement, and must be used in conjunction with Animal Component-Free Cell Attachment Substrate. STEMdiff™ Endothelial Expansion Medium is optimized for the expansion of hPSC-derived endothelial cells and for the maintenance of endothelial marker expression. For passaging, Animal Component-Free Cell Dissociation Kit is required. NOTE: Heparin Solution is required for preparation of complete STEMdiff™ Endothelial Expansion Medium. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').