ContactUsLinkComponent
ReproTeSR™ Medium for Reprogramming (2-Component)
ReproTeSR™ Medium for Reprogramming (2-Component)
Catalog # MSPP-05926
CAS Number:  
undefined
ReproTeSR™ Medium for Reprogramming (2-Component)
Catalog # MSPP-05926
Supplier Number:  05926
CAS Number:  
Specifications
  • Description:
    ReproTeSR™ medium for reprogramming (2-component)
  • Size:
    500 ml
  • Cat. No.:
    MSPP-05926
Specifications
About this item
Reprogramming medium for human iPS cell induction.
ReproTeSR™ (2-Component) is a complete and xeno-free reprogramming medium developed for generating human induced pluripotent stem (iPS) cells from fibroblasts, urine-derived cells, and blood-derived cells such as CD34+ or erythroid precursor cells under feeder-free conditions. ReproTeSR™ is also available as part of an integrated set of tools that are optimized for reprogramming blood-derived cell types expanded in vitro from peripheral blood, such as CD34+ cells (CD34+ Progenitor Reprogramming Kit) or erythroid cells (Erythroid Progenitor Reprogramming Kit).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').