ContactUsLinkComponent
PneumaCult™-ALI-S Medium
PneumaCult™-ALI-S Medium
Catalog # MSPP-05050
CAS Number:  
undefined
PneumaCult™-ALI-S Medium
Catalog # MSPP-05050
Supplier Number:  05050
CAS Number:  
Specifications
  • Description:
    PneumaCult™ ALI-S medium
  • Size:
    1 kit
  • Cat. No.:
    MSPP-05050
Specifications
About this item
Serum- and BPE-free medium for human small airway epithelial cells cultured at the air-liquid interface.
PneumaCult™-ALI-S Medium is a serum- and BPE-free medium for the culture of human small airway epithelial cells at the air-liquid interface (ALI). Small airway epithelial cells cultured in PneumaCult™-ALI-S Medium undergo extensive mucociliary differentiation to form a cuboidal epithelium that exhibits morphological and functional characteristics similar to those of the human small airway in vivo. Together, PneumaCult™-ALI-S Medium and PneumaCult™-Ex Plus Medium constitute a fully integrated BPE-free culture system for in vitro human small airway modeling. This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').