ContactUsLinkComponent
MethoCult™ Starter Kit for H4034 Optimum
MethoCult™ Starter Kit for H4034 Optimum
Catalog # MSPP-04064
CAS Number:  
undefined
MethoCult™ Starter Kit for H4034 Optimum
Catalog # MSPP-04064
Supplier Number:  04064
CAS Number:  
Specifications
  • Description:
    Starter kit for MethoCult™ H4034 optimum
  • Size:
    1 kit
  • Cat. No.:
    MSPP-04064
Specifications
About this item
Complete kit for hematopoietic CFU assays.
The Starter Kit for MethoCult™ H4034 Optimum (MethoCult™ GF H4034) is recommended for laboratories in the initial stages of establishing procedures for assessing human hematopoietic progenitor cells using colony-forming unit (CFU) assays. These products support the growth of clonogenic hematopoietic progenitor cells from human bone marrow, peripheral blood, cord blood, leukapheresis products and purified progenitor-enriched cells. H4034 supports the growth of erythroid progenitors (BFU-E and CFU-E), granulocyte-macrophage progenitors (CFU-GM, CFU-G and CFU-M) and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitors (CFU-GEMM). Each kit contains instructional materials in addition to all reagents and materials necessary to perform 24 duplicate assays.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').