ClonaCell™-HY Medium
Catalog # MSPP-03803
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Description:ClonaCell™ HY medium C
- Size:100 ml
- Cat. No.:MSPP-03803
Specifications
About this item
Hybridoma fusion recovery medium (serum-containing).
ClonaCell™-HY Medium C is a . Serum-containing liquid medium used to promote hybridoma viability and recovery after the fusion process (PEG-based or electrofusion) and prior to hypoxanthine, aminopterin, thymidine (HAT) selection. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').