ContactUsLinkComponent
EpiCult™-B Mouse Medium Kit
EpiCult™-B Mouse Medium Kit
Catalog # MSPP-05610
CAS Number:  
undefined
EpiCult™-B Mouse Medium Kit
Catalog # MSPP-05610
Supplier Number:  05610
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    EpiCult™-B mouse medium kit
  • Size:
    500 ml
  • Cat. No.:
    MSPP-05610
Specifications
About this item
For culture and evaluation of mouse mammary epithelial cells.
EpiCult™-B (Mouse) is a serum-free liquid culture medium optimized for the culture of mouse mammary luminal and myoepithelial cells. It is ideal for the culture and evaluation of mouse mammary epithelial progenitors in the mammary colony-forming unit assay when used in conjunction with an irradiated feeder layer such as NIH 3T3 cells. This medium is also used for enzymatic dissociation of mouse mammary tissue when supplemented with collagenase and hyaluronidase. Addition of human recombinant EGF, human recombinant bFGF, and heparin solution is required for culturing cells.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').