ContactUsLinkComponent
STEMdiff™ Hematopoietic Kit
  MSPP-05310
 :  
undefined
STEMdiff™ Hematopoietic Kit
  MSPP-05310
 :  05310
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    STEMdiff™ hematopoietic kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-05310
For differentiation of human PSCs into the hematopoietic lineage.
Generate functional hematopoietic progenitor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells with STEMdiff™ Hematopoietic Kit. Using a simple and reproducible protocol in serum- and feeder-free conditions, you can produce hematopoietic progenitor cells expressing CD34, CD45, and CD43. The robust, two-stage protocol initially induces cells toward the mesoderm before further differentiating them into hematopoietic progenitor cells. After 12 days, a population of hematopoietic cells containing 25 to 65% (average 43%) CD34+CD45+ progenitor cells can be harvested, including progenitor cells that have the capacity to form functional hematopoietic colonies in the colony-forming unit (CFU) assay.
STEMdiff™ hematopoietic kit has been optimized for differentiation of human pluripotent cells maintained in:
mTeSR™1
mTeSR™ plus
TeSR™-E8™
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').