Orders LinkContactUsLinkComponent
Draft Shields for Equipments
Draft Shields for Equipments
Supplier:   Mettler Toledo
Draft Shields for Equipments
Supplier:  Mettler Toledo
Certificates
Draft Shields for Equipments
Draft Shields for Equipments
2 Options Available Below
Product Details & Documents
The draft shield minimizes air turbulence around your balance, enabling results to be delivered faster.
  • Faster stabilization time
  • Speed up your weighing processes
Product Family Options
Product Information
  • Height
    Availability
    Price
  • 97044-172
    Draft shield with sliding doors, 1 mg
    Draft shield with sliding doors, 1 mg
    168 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,130.00
    $1,130.00
    Each
       $1,130.00
    Supplier Number: 12122405
  • 97044-174
    Draft shield with sliding doors, 0.1 mg
    Draft shield with sliding doors, 0.1 mg
    236 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,250.00
    $1,250.00
    Each
       $1,250.00
    Supplier Number: 12122404