OIML Class M1 Standard Line Weights
Supplier: Mettler Toledo
Certificates
About this item
Single M1 OIML class sheet and knob weights in a plastic box for standard environments.
- Affordable quality weights for regular routine testing without the need for traceability
- No calibration certificate
- 304 Stainless steel
- <0.8 Susceptibility X
- 7950 (±140) kg/m³ density ρ
Adjusting cavity: Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.
Frequently Bought Together