ContactUsLinkComponent
TexQ® Ready-to-Use (RTU) Disinfectant Cleaner
Catalog # 77394-850
Supplier:  Texwipe
undefined
TexQ® Ready-to-Use (RTU) Disinfectant Cleaner
Catalog # 77394-850
Supplier:  Texwipe
Supplier Number:  TX652
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    TexQ® RTU disinfectant cleaner
  • Size:
    1 gallon bottle
  • Cat. No.:
    77394-850
  • Sterility:
    Sterile
Specifications
About this item
TX650 TexQ® disinfectant is an EPA-registered, one-step disinfectant and cleaner that is highly effective against a wide spectrum of bacteria, viruses, and fungi. It is fragrance-free, dye-free, and 0.2 micron filtered. Gamma Irradiated and sterile validated according to AAMI guidelines.
  • Kill claims provide assurance of disinfection efficacy against most common cleanroom bacteria, viruses, and fungi
  • RTU solutions are gamma irradiated to a SAL of 10⁻⁶
  • Compliant with aseptic environment requirements per USP <797> and USP <800>
  • Excellent cleaning and disinfecting properties for uniform and complete surface disinfection
  • Cleans and disinfects in one step, saving costs in time and labor
  • Free of dyes and fragrance, no respiratory irritation
  • Easy documentation and usage control, record the date opened and operator initials
Effective against a broad spectrum of bacteria, viruses, and fungi including MRSA, Pseudomonas, H5N1, HIV-1, SARS-CoV-2 (COVID-19 virus) at one, two, or ten minutes contact time. This disinfectant cleaner inhibits the growth of mold and mildew and their odors when used as directed. Recommended for everyday use as part of a disinfectant rotation program.
Compatible with glass, laminated surfaces, metal, stainless steel, glazed porcelain, glazed ceramic, sealed granite, sealed marble, plastic (such as polycarbonate, polyvinylchloride, polystyrene, or polypropylene), sealed limestone, sealed slate, sealed stone, aluminum, sealed terra cotta, sealed terrazzo, chrome and vinyl, Plexiglas®, vanity tops, enameled surfaces, painted/finished woodwork, Formica®, vinyl and plastic upholstery, washable wallpaper, windows and mirrors.
Packaging: Packed in a one gallon bottle, 4 bottles per case.