Human Recombinant RhoA Mutant (S188A, His tag) (from E. coli)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant human RhoA (S188A) (1-190) was expressed in E. coli cells using an N-terminal His tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:ARHA|RHOH12|ARH12|RHO12
- Protein/Peptide Name:RHOA
- Molecular Weight:23 kDa
- Formulation:Supplied as sterile 50 mM sodium phosphate, pH 7.5, 300 mM NaCl, 50 - 200 mM imidazole, 0 - 0.1 mM PMSF, 0.2 - 1.0 mM DTT, 10 - 25% glycerol.
- Tested Applications:WB, Kinase assay
Specifications
Frequently Bought Together