Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant PKC gamma (GST tag), Active (from Baculovirus-Insect Cells)
New Product
 
Human Recombinant PKC gamma (GST tag), Active (from Baculovirus-Insect Cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    PKCG|SCA14|PKCC|PKC-gamma
  • Protein/Peptide Name:
    PRKCG
  • Molecular Weight:
    ~105 kDa
  • Formulation:
    Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol

 

Frequently Bought Together