Order Entry
United States
ContactUsLinkComponent
Human Recombinant PNMT (GST tag), Active (from Baculovirus-Insect Cells)
Human Recombinant PNMT (GST tag), Active (from Baculovirus-Insect Cells)
Supplier:  Sino Biological
New Product
Certificates
Human Recombinant PNMT (GST tag), Active (from Baculovirus-Insect Cells)
Supplier:  Sino Biological
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    PENT|PNMTase
  • Protein/Peptide Name:
    PNMT
  • Molecular Weight:
    57 kDa
  • Formulation:
    Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
  • Tested Applications:
    Methyltransferase assay

Specifications

Related Information

Frequently Bought Together