Human Recombinant EZH2 (Y641S, GST tag) (from Baculovirus (Insect cells))
Recombinant full length human EZH2 (Y641S) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:KMT6|EZH2b|WVS|ENX1|ENX-1|KMT6A|WVS2|EZH1
- Protein/Peptide Name:EZH2
- Molecular Weight:130 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
- Tested Applications:Methyltransferase Assay
Frequently Bought Together