Human Recombinant IRS1 (C-Term, GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant Human IRS1 (600 to 1245) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:HIRS-1
- Protein/Peptide Name:IRS1
- Molecular Weight:~118 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Specifications
Related Information
Frequently Bought Together