Order Entry
United States
ContactUsLinkComponent
CHOSEN™ Non-Contact Infrared Thermometer
CHOSEN™ Non-Contact Infrared Thermometer
Catalog # 77530-066
undefined
CHOSEN™ Non-Contact Infrared Thermometer
Catalog # 77530-066
Supplier Number:  FTH800

Specifications

  • Description:
    Non-contact infrared thermometer
  • Cat. No.:
    77530-066
  • Range:
    93.2...109.4 °F

Specifications

About this item

Reliable, fast results in one second.

  • Reliable and fast results
  • Advanced infrared technology provides quick and accurate measurements in one second
  • Non-contact provides safer measurements
  • No measuring sensor cover is needed
  • Minimizes the risk of spread of disease and cross-contamination
  • Eliminates the need for cleaning between each person
  • Easy to use
  • User-friendly design offers comfortable grip
  • Storage for up to 50 measurement memories
  • Switchable between °F/°C and beep/silent options
  • Five-year shelf life

Features wide body temperature measuring range between 93.2 to 109.4°. Offers reliable results in a wide environmental temperature range at 50 to 104 °F.

Hold the non-contact thermometer 0.4 to 2" (1 to 5 cm) away from the center of forehead to obtain measurement. Easy to read LCD display includes three backlight colors and features red high fever alert above 101.3°.

Ordering information: Fast shipping.
Delivery: 2 AAA batteries included.