Order Entry
United States
ContactUsLinkComponent
 
Wire Stand for Autoclave Bags
  104026-718
undefined
Wire Stand for Autoclave Bags
  104026-718
 :  A3004-WR

 

  • Description:
    Wire stand for 8.5×11" autoclave bags, 1/pack
  • Cat. No.:
    104026-718

 

 

Crafted from robust and corrosion-resistant wire, this stand is built to withstand the rigors of daily use in medical and laboratory environments. The high-quality materials ensure longevity and reliability for prolonged usage.

  • Suitable for for 8.5×11" autoclave bags
  • Autoclave temperature: 121 °C; maximum 136 °C