Benchtop Disposal Set
Catalog # 104027-712
Supplier: Alkali Scientific
Specifications
- Description:Benchtop disposal set, 5 ml, includes metal stand (5×3×9") and 400 red biohazard bags (8.5×11")
- Cat. No.:104027-712
Specifications
About this item
The perfect benchtop disposal container for tips, tubes, gloves, plates and more.
- Complete kit - 400 bags and metal stand
- Compact - only 3×5" footprint
- Sturdy - steel construction
- Autoclavable
Delivery information: Supplied with metal stand (5×3×9") and 400 red biohazard bags (8.5×11").